missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Poly-D-Lysine/Laminin Multiwell Plate

Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology

Brand:  Corning™ 354595

Code : 67

Additional Details : Weight : 0.55000kg

 View more versions of this product

Product Code. 10594841

  • £196.51 / Pack of 5


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Alert:

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.


Description

Description

  • Matrix components are applied as an even, optically clear coating
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells

Applications:

For adhesion, growth and differentiation of neuronal and transfected cell lines

Specifications

Specifications

6-Well Poly-D-Lysine/Laminin Multiwell Microplate
9.6cm2
6 well
Poly-D-Lysine and Laminin
2° to 8°C
Clear
Round
Clear
Enhancement of neuronal cell attachment to plastic and glass, promotion of neurite outgrowth, culture of glial cells as a feeder layer for neurons, construction of neural cell model systems to study CNS function, development and diseases
Polystyrene
Non-sterile
ECM/Attachment Factors
With Lid
Videos

For research use only.