missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ Poly-D-Lysine Culture Dish
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Brand: Corning™ 356469
Code : 66
Additional Details : Weight : 1.13000kg
Description
- All lots tested and found negative for bacteria and fungi
- Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
- Coated with 0.2 micron filtered PDL in a cleanroom
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
Clear | |
Polystyrene | |
Clear | |
Polystyrene | |
Poly-D-Lysine | |
Lid | |
100 mm |
Closure Included | |
Round | |
58.1 sq. cm | |
Round | |
Cell Culture Dish | |
40/cs. |