missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Matrigel™ Matrix Multiwell Plates

Soluble basement membrane extracted from Engelbreth-Holm-Swarm mouse tumor forms a gel when reconstituted at room temperature

£180.00 - £556.56

Specifications

Material Polystyrene
Color Clear
Sterility Non-sterile
Source EHS Mouse Tumor
Well Shape Round
View More Specs

Products 6
Product Code Brand No. of Wells Price Quantity & Availability  
Product Code Brand No. of Wells Price Quantity & Availability  
10681274 View Documents Corning™
354671
6 (for ES culture) £180.00
Pack of 5 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
10203901 View Documents Corning™
354503
12 £435.00
Pack of 2 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
10142561 View Documents Corning™
354508
48 £440.00
Pack of 2 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
10556011 View Documents Corning™
354603
6 £221.00
Pack of 5 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
10020901 View Documents Corning™
354510
6 (for hepatocytes) £556.56
Pack of 5 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
10546011 View Documents Corning™
354605
24 £212.53
Pack of 5 View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Corning Matrigel Basement Membrane Matrix is a solubilized tissue basement membrane preparation containing laminin, collagen type IV, heparan sulfate proteoglycan and entactin. Corning Matrigel Matrix is useful as a substrate for growth and differentiation of many normal and transformed cell types, such as epithelial, endothelial, muscle and neuronal cells. Corning BioCoat Thin Layer Matrigel cultureware contain a dried thin layer of Corning Matrigel Basement Membrane Matrix. Rehydration results in the formation of a thin “basement membrane” gel layer on the cultureware surface. Specific applications for Thin Layer Matrigel cellware include culturing of primary contractile skeletal myotubes, neuronal cell morphological differentiation, studies of vascular smooth muscle cell interaction with the extracellular matrix and the effects of the extracellular matrix on the function and morphology of lung alveolar, and bile ductular epithelial cells.

  • Especially suited for culture of polarized cells, such as epithelial cells, and promotes differentiation of hepatocytes, mammary epithelial cells and smooth muscle cells
  • Promotes capillary tubule formation of endothelial cells and neurite regeneration
  • Can be used for angiogenesis assays
  • Tested for the ability to promote neurite outgrowth from chick dorsal root ganglia (in the absence of NGF)

Specifications

Specifications

Polystyrene
Non-sterile
Round
Clear
EHS Mouse Tumor

For research use only