Learn More
Corning™ BioCoat™ Laminin Rectangular Canted Neck Cell Culture Flask w/Plug Seal Cap
Tested for their ability to initiate differentiation (neurite outgrowth) of NG-108 rat glioma/mouse and human neuroblastoma cells
Brand: Corning™ 354533
Code : 67
Additional Details : Weight : 0.33000kg
Description
- Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
- Matrix components are applied as an even, optically clear coating
- All lots tested and found negative for bacteria and fungi
- Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
These products are for research use only and are not intended for diagnostic purposes.
For adhesion, growth and differentiation of neuronal and transfected cell lines
Specifications
2.36 oz. | |
70 mL | |
Canted | |
Laminin | |
25cm2 |
Phenolic | |
For adhesion, growth and differentiation of neuronal and transfected cell lines | |
Rectangular | |
Sterile |