missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ Fibronectin TC-treated Culture Dish
Matrix components are applied as an even, optically clear coating
£199.02 - £266.43
Specifications
Bottom Shape | Round |
---|---|
Color | Clear |
Material | Polystyrene |
Shape | Round |
Type | Cell Culture Dish |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10747351
|
Corning™
354451 |
100 mm |
£225.76
Pack of 10 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
10080741
|
Corning™
354403 |
60 mm |
£213.52
Pack of 20 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
10214091
|
Corning™
354457 |
35 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10152801
|
Corning™
354552 |
150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
- Tested for their ability to promote attachment and spreading of BHK-21 hamster kidney cells
- All lots tested and found negative for bacteria and fungi
- Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
The human plasma used in preparing this product was tested and found negative for HBsAg and HIV antibody.
For adhesion, growth and differentiation of neuronal and transfected cell lines
Order Info
Bulk quantities available; please contact your Customer Service Representative.
Specifications
Round | |
Polystyrene | |
Cell Culture Dish |
Clear | |
Round |