Learn More
Corning™ BioCoat™ 384-Well, Fibronectin-Treated, U-Shaped-Bottom Microplate
Color:Clear; Sterility:Non-sterile; Lid:With Lid; Type: Optical Glass Base
Brand: Corning™ 4585
Code : 67
Additional Details : Weight : 2.45000kg
Description
High optical quality, glass bottom black microplates are ideal for performing high-content cell-based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput, and is ideal for cell growth.
- High optical quality and scratch resistant glass
- Glass bottom thickness of 200 μm is well suited for imaging microscopy.
- Bottom flatness <50 μm to ensure planarity for imaging devices
- Low background fluorescence and minimal cross-talk provide the highest possible optical quality for cell-based assays.
Order Info
Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates
Specifications
Standard | |
2°C to 8°C | |
16 x 24 | |
Polystyrene | |
Non-sterile | |
Round Bottom (U-well) | |
10/Cs. | |
Human Fibronectin |
With Lid | |
0.32 cm2 | |
Clear | |
384 | |
Fibronectin | |
Promotion of cell attachment and spreading, rapid expansion of cell populations, increased proliferation of PC12 cells, studies of effects of collagen IV on cell behavior, cell adhesion assays | |
1/Pk. | |
Clear |