Learn More
Corning™ BioCoat™ Fibronectin, Human Cell Culture Insert
Tested for their ability to promote attachment and spreading of BHK-21 hamster kidney cells
Brand: Corning™ 354445
Code : 67
Additional Details : Weight : 0.23000kg
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Description
- Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology Supplied with preapplied extracellular matrices, facilitating cell adaption to in vitro cultivation
- Microporous membranes permit diffusion of ions, low-molecular-weight proteins and other substances to both apical and basolateral cell surfaces
- Low pore-density, track-etched polyethylene teraphthalate (PET) membranes for convenient viewing of living cells with a phase-contrast microscope
- Have low-binding properties that minimize small molecule loss
The human plasma used in preparing this product was tested and found negative for HBsAg and HIV antibody.
Order Info
Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates
Specifications
PET | |
Membrane Insert | |
2° to 8°C |
0.4 μm | |
24 Well Plate |