Learn More
Corning™ BioCoat™ Collagen IV TC-Treated Culture Dishes
Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
Brand: Corning™ 354459
Code : 67
Additional Details : Weight : 0.15000kg
Description
Microporous membranes permit diffusion of ions, low-molecular-weight proteins and other substances to both apical and basolateral cell surfaces, simulating the natural environment of cells in vivo.
- Source: Mouse
- TC-treated
- Nonsterile
For adhesion, growth and differentiation of neuronal and transfected cell lines
Specifications
Round | |
Round | |
Cell Culture Dish |
Polystyrene | |
Collagen Type IV | |
35 mm |