missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ 96-Well, Collagen Type I-Treated, Flat-Bottom Microplate
GREENER_CHOICE

Catalog Number 10591721
Click to view available options
Packaging:
5/Cs
5/Pk., 50/Cs.
80/Cs
This item is not returnable. View return policy
This item is not returnable. View return policy

Color: White, Clear Bottom; Sterility: Sterile; Lid: With Lid

Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.

Applications include:

  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced serum culture
  • Cell adhesion assays
  • Improving survival of primary cells in culture

Source

  • Rat tail tendon
  • Quality Control

    • Tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
    • Tested and found negative for bacteria and fungi
    • Collagen I purity >90% by SDS-PAGE

    Storage

  • 4°C to 30°C under dry conditions.
  • TRUSTED_SUSTAINABILITY
    Certifications/Compliance ANSI, SBS
    Format Standard
    Lid With Lid
    Content And Storage 4°C to 30°C
    Culture Area 0.32 cm2
    Array 8 x 12
    Packaging 5/Cs
    Color White, Clear Bottom
    Material Polystyrene
    No. of Wells 96
    Sterility Sterile
    Surface Treatment Collagen Type I
    Well Shape Round, Flat-bottom
    For Use With (Application) Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture
    No. per Case 5/Cs.
    No. per Pack 5/Pk.
    Quantity 5/Cs.
    Source Rat Tail
    Well Color Clear
    Show More Show Less
    missing translation for 'provideContentCorrection'

    missing translation for 'productTitle'