Learn More
Corning™ BioCoat™ #1 German Glass Coverslips, Round
Coated with a variety of extracellular matrix substrates. Corning™ BioiCoat™ No. 1 German Glass Coverslips provide an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures.
Brand: Corning™ 354088
Code : 67
Additional Details : Weight : 0.45360kg
Description
Corning™ BioCoat™ German Glass coverslips with surface treatment (see individual Specification tables for coating). Corning BioCoat products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
Shelf life: ≥ 3 months from date of shipment
Specifications
Slide Coverslip | |
No. 1 German Glass | |
Fluorescence imaging of live cells, confocal microscopy, high resolution and inverted microscopy, phase contrast microscopy | |
60 |
Fibronectin | |
0.866 in. | |
22 mm | |
60 |