Learn More
Corning™ Bar Coded Storage Tubes:2D Bar Coded
For enhanced sample storage, identification and retrieval
Brand:  Corning™ 8511
Code : 66
Additional Details : Weight : 1.39000kg
 View more versions of this product
Product Code. 11764955
  • £1990.71 / Pack of 38400
Estimated Shipment
27-02-2018
Add to basket
Specifications
Specifications
Polypropylene
Orange
No
Plug-seal
Bulk
Plug Caps for Round Tubes
Plug Seal
Polypropylene
384 2D bar-coded round tubes, enhanced sample storage, identification and retrieval
14mm
2D Barcoded
Videos