Learn More
Corning™ 96-Well Cell Culture Multiwell Plates
Designed for use in most thermal cyclers and offer a rigidity that makes them ideal for high-throughput (HT) automation.
Brand:  Corning™ 9520
Code : 66
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 12706005
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
Sold 100 per case.
Specifications
Specifications
96-Well Cell Culture Multiwell Plates
100/Cs.
Non-Treated
Polypropylene
Non-sterile
Videos