missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 50mL Easy ID Color Strip Polypropylene Centrifuge Tube
Easily identify samples contained in clear, color strip polypropylene tubes that eliminate sticker labeling
£127.59 - £142.04
Specifications
Closure Color | Orange |
---|---|
Disposable | Single-use |
Material | Polypropylene |
For Use With | Safe for air shipping |
Length (Metric) | 114.4 mm |
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
15407471
|
Corning™
431526 |
Green band |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15427471
|
Corning™
431527 |
Orange band |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15497461
|
Corning™
431525 |
Hazardous materials band |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15447471
|
Corning™
431528 |
Blue band |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Conical bottom
- Plug seal
- Maximum RCF is 15,500xg
- Sterile, nonpyrogenic, and certified RNase-/DNase-free
- 95 k PA (14 psi) pressure tested
- Recommended working temperature: 0-40°C
- Heat distortion point: 121°C
Specifications
Orange | |
Polypropylene | |
114.4 mm | |
Yes |
Single-use | |
Safe for air shipping | |
29.1 mm | |
15,500 x g |