missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 1536-Well, Cell Culture-Treated, Flat-Bottom Microplate
Click to view available options
Packaging:
10/Pk., 50/Cs.
Description
- Flat bottom wells are compatible with automated dispensers including the PerkinElmer FlexDrop™
- Working volume of up to 10μL
- Available untreated, with nonbinding surface (NBS™), or tissue culture treated
- NBS microplates are ideal for homogeneous assays with “sticky” enzymes or for reducing nonspecific binding to microplate wells
Generic Barcode
- Ensures compatibility with most bar code scanning and software systems
- To avoid mix-ups, the surface treatment of the microplate wells is easily identified in the human readable portion of the bar code (NT = not treated; TC = tissue culture treated; NB = nonbinding surface)
- Labels applied to all four sides of the plate
- No sequence number duplicates
Specifications
Barcode | No Barcode |
Format | Standard |
Lid | With Lid |
Culture Area | 0.03 cm2 |
Array | 32 x 48 |
Packaging | 10/Pk., 50/Cs. |
Color | Black |
Product Type | 1536-Well Assay Microplate |
Material | Polystyrene |
No. of Wells | 1536 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'