missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Copper tubing, 1.59mm (0.0626in) OD, 0.876mm (0.0345in) ID, 99.9% (metals basis)

Brand:  Alfa Aesar™ 013474.RM

Code : NEW

Additional Details : CAS Number : 7440-50-8 Weight : 0.01000kg

Quantity 90cm

 View more versions of this product

Product Code. 11356838

  • £7.00 / 90cm


Estimated Shipment date
from Supplier 24-05-2018
Add to basket

Specifications

Chemical Identifiers

Copper tubing
7440-50-8
OD: 1.59mm (0.0626 in.) ID: 0.876mm (0.0345 in.)
MFCD00010965

Specifications

Copper tubing
OD: 1.59mm (0.0626 in.) ID: 0.876mm (0.0345 in.)
99.9%
MFCD00010965
90cm
7440-50-8
(metals basis)
Odorless
Videos
SDS