Learn More
Thermo Scientific Alfa Aesar Copper tubing, 1.59mm (0.0626in) OD, 0.876mm (0.0345in) ID, 99.9% (metals basis)
Brand: Thermo Scientific Alfa Aesar 013474.RN
Copper tubing, 1.59mm (0.0626in) OD, 0.876mm (0.0345in) ID, 99.9% (metals basis)
Code : NEW
Additional Details : CAS Number : 7440-50-8 Weight : 0.01000kg