missing translation for 'onlineSavingsMsg'
Learn More

Copper sputtering target, 76.2mm (3.0in) dia x 6.35mm (0.250in) thick, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 15464785
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 Ea.
CAS 7440-50-8
Odor Odorless
Molecular Formula Cu
MDL Number MFCD00010965
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES [Cu]
IUPAC Name copper
Molecular Weight (g/mol) 63.55
PubChem CID 23978
ChEBI CHEBI:30052
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Copper sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'