missing translation for 'onlineSavingsMsg'
Learn More

Copper sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.995% (metals basis), Thermo Scientific™

Catalog Number 15494885
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 7440-50-8
Molecular Formula Cu
MDL Number MFCD00010965
Quantity 1 Ea.
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES [Cu]
IUPAC Name copper
Molecular Weight (g/mol) 63.55
PubChem CID 23978
ChEBI CHEBI:30052
Percent Purity 99.995%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Copper sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'