missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.995% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 Ea.
Description
Sputtering targets commonly used to coat electronic components and electronic instruments. Due to its high electrical conductivity, copper is becoming increasingly popular for use in large-format, high-resolution TFT-LCD television sets. Copper is used as the electrode layer in thin-film transistors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 1 Ea. |
CAS | 7440-50-8 |
Odor | Odorless |
Molecular Formula | Cu |
MDL Number | MFCD00010965 |
Synonym | cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer |
Solubility Information | Insoluble in water. |
InChI Key | RYGMFSIKBFXOCR-UHFFFAOYSA-N |
SMILES | [Cu] |
IUPAC Name | copper |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'