missing translation for 'onlineSavingsMsg'
Learn More

Copper Solution for ICP, 1g/L, HNO3 Matrix, Fisher Chemical™

Catalog Number p-8014468
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 1g/L
Quantity 100 mL
Solution Contains HNO3
compliance-icons
Product Identifier
  • Single-element Standard Solution for ICP. Copper (Cu) 1000mg/l in HNO3 2%
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'