missing translation for 'onlineSavingsMsg'
Learn More

Copper Shot, 0.6-0.8mm (0.02-0.03 in.), 99.5% (Metals basis), Alfa Aesar™

Brand:  Alfa Aesar 044212.36

Code : NEW

Additional Details : CAS Number : 7440-50-8 Weight : 0.50000kg

Quantity 500g

 View more versions of this product

Product Code. 11379718

  • £40.00 / 500g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

Copper
Shot, 0.6-0.8mm (0.02-0.03 in.)
MFCD00010965
7440-50-8
99.5% (Metals basis)

Specifications

Copper
7440-50-8
Shot, 0.6-0.8mm (0.02-0.03 in.)
MFCD00010965
Copper colored
500g
Copper colored shot
99.5% (Metals basis)
Odorless
Videos
SDS