missing translation for 'onlineSavingsMsg'
Learn More

Copper powder, spherical, -170+270 mesh, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7059878
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]
This item is not returnable. View return policy
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Red
UN Number UN3089
Quantity 100 g
Percent Purity 99.9%
Mesh Size −170+270 Mesh
Odor Odorless
Form Sphere
Assay (metals basis)
Chemical Name or Material Copper powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'