missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Copper plate, Oxygen-Free High Conductivity (OFHC), alloy 101, 3.18mm (0.125 in.) thick

Brand:  Alfa Aesar™ 045224.GT

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 150x150mm

 View more versions of this product

Product Code. 15493955

  • £90.63 / Each


Estimated Shipment date
from Supplier 25-04-2018
Add to basket

Specifications

Chemical Identifiers

Copper plate, Alloy 101
Oxygen-Free High Conductivity (OFHC), 3.18mm (0.125 in.) thick
MFCD00010965

Specifications

Copper plate, Alloy 101
Oxygen-Free High Conductivity (OFHC), 3.18mm (0.125 in.) thick
MFCD00010965
150x150mm
Plate
Videos
SDS