Learn More
Copper plate, Oxygen-Free High Conductivity (OFHC), alloy 101, 2.4mm (0.093in) thick, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 045210.GT
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Copper plate, Alloy 101 | |
MFCD00010965 | |
Plate |
Oxygen-Free High Conductivity (OFHC), 2.4mm (0.093 in.) thick | |
150 x 150 mm |
RUO – Research Use Only