missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Copper Nickel wire, 0.5mm (0.02 in.) dia.

Brand:  Alfa Aesar™ 043272.H9

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11309388

  • £163.10 / 500m


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Specifications

Cu:Ni 55:45 wt%
≈1.75g/m
0.5mm (0.02 in.) dia.
MFCD00801098
Copper Nickel wire
500m
Wire
Videos
SDS