missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) tungsten oxide, 99.5% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
Copper(II) tungsten oxide nanopowder is used for supercapacitor applications. Nano-sized CuWO4 thin films have been fabricated by radio-frequency (R.F.) sputtering deposition, and are used as positive electrode with both LiClO4 liquid electrolyte and LiPON solid electrolyte in rechargeable lithium batteries. Thin films of copper tungstate is used for nitric oxide sensing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 50 g |
Merck Index | 14,2657 |
Solubility Information | Insoluble in water. |
Formula Weight | 311.38 |
Percent Purity | 99.5% |
Odor | Odorless |
Assay | (metals basis) |
Chemical Name or Material | Copper(II) tungsten oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'