missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) Trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals
36.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Blue to White or Gray |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Linear Formula | (CF3SO3)2Cu |
Quantity | 5 g |
Fieser | 07,76; 08,126; 10,110; 14,102 |
Formula Weight | 361.69 |
Percent Purity | 98% |
Physical Form | Crystalline Powder |
Chemical Name or Material | Copper(II) trifluoromethanesulfonate |
Safety and Handling
Product Identifier
- Copper(II) trifluoromethanesulfonate
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'