missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) sulfide, ultra dry, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7049415
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
1317-40-4
CuS
95.61
MFCD00016066
OMZSGWSJDCOLKM-UHFFFAOYSA-N
copper ii sulfide, copper monosulfide, cupric sulfide, copper sulfide, copper blue, copper sulphide, oil blue, covellite, copper 2+ sulfide, horace vernet's blue
14831
[S--].[Cu++]
This item is not returnable. View return policy
1317-40-4
CuS
95.61
MFCD00016066
OMZSGWSJDCOLKM-UHFFFAOYSA-N
copper ii sulfide, copper monosulfide, cupric sulfide, copper sulfide, copper blue, copper sulphide, oil blue, covellite, copper 2+ sulfide, horace vernet's blue
14831
[S--].[Cu++]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Melting Point 220°C (decomposition)
Physical Form Powder
Quantity 10 g
Merck Index 14,2965
IUPAC Name sulfanylidenecopper
Formula Weight 95.6
Percent Purity 99.98%
Odor Odorless
Packaging Ampule
Refractive Index 1.45
Sensitivity Air and moisture sensitive
Density 3.9 g/mL to 4.6 g/mL
Assay (metals basis)
Chemical Name or Material Copper(II) sulfide, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'