missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) sulfate pentahydrate, Puriss., suitable for, meets analytical spec. of BP, FCC, Ph. Eur., USP, Honeywell Fluka™
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
2.5 kg
6 x 500 g
25 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 100 g |
Linear Formula | CuSO4 · 5H2O |
UN Number | 3077 |
IUPAC Name | copper;sulfate;pentahydrate |
Formula Weight | 249.69g/mol |
Grade | puriss. |
Vapor Pressure | 7.3mmHg (25°C) |
Chemical Name or Material | Copper(Ii) Sulfate Pentahydrate, Meets analytical specification of Ph. Eur.BP, USP, FCC |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'