missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) Sulfate Pentahydrate, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
1 kg
3 kg
5 kg
Specifications
Melting Point | 110°C |
Color | Blue |
pH | 3.5 to 4.5 |
Physical Form | Solid |
Quantity | 250 g |
Formula Weight | 249.68g/mol |
Packaging | Plastic powder jar |
Vapor Pressure | 7.3mmHg 25 |
Safety and Handling
Product Identifier
- COPPER (II) SULFATE PENTAHYDRATE
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
- P273-Avoid release to the environment.
- MIXTURE LIST-Contains : Copper (II) sulfatepentahydrate (1:1:5), Cupric sulfate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'