missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) selenite hydrate, 97%, Alfa Aesar™

Brand:  Alfa Aesar 040221.18

Code : NEW

Additional Details : CAS Number : 10214-40-1 Weight : 0.05000kg Transport : UN number : 2630 Chem class : 2 Pack group : I

Quantity 50g

 View more versions of this product

Product Code. 11336499

  • £82.80 / 50g


Estimated Shipment date
from Supplier 14-02-2018
Add to basket

Specifications

Chemical Identifiers

Copper(II) selenite hydrate
Crystalline Powder
CuO3Se·xH2O
MFCD00010973
10214-40-1
97%
190.50 (Anhydrous)

Specifications

Copper(II) selenite hydrate
Crystalline Powder
CuO3Se·xH2O
190.50 (Anhydrous)
Blue
10214-40-1
97%
Blue crystalline powder
MFCD00010973
50g
Videos
SDS