missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) pyrophosphate hydrate, P{2}O{7} 43.5-48.5%, Thermo Scientific Chemicals

Catalog Number p-7059008
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable. View return policy
16570-28-8
Cu2O7P2
301.03
MFCD00150225
PEVJCYPAFCUXEZ-UHFFFAOYSA-J
dicopper(2+) (phosphonatooxy)phosphonate
[Cu++].[Cu++].[O-]P([O-])(=O)OP([O-])([O-])=O
This item is not returnable. View return policy
16570-28-8
Cu2O7P2
301.03
MFCD00150225
PEVJCYPAFCUXEZ-UHFFFAOYSA-J
dicopper(2+) (phosphonatooxy)phosphonate
[Cu++].[Cu++].[O-]P([O-])(=O)OP([O-])([O-])=O

Copper(II) pyrophosphate hydrate is used in printed circuit boards. It catalyzes the dimerization of isobutylene to diisobutylene. It is involved in the electrodeposition of copper-zinc alloys based electrolytes. Further, it is used in animal feed as a supplement. In addition to this, it is used in plating of copper on plastics, aluminum, and zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Blue
Physical Form Powder
Quantity 1 kg
Solubility Information Insoluble in water.
Formula Weight 301.02 (Anhydrous)
Concentration or Composition (by Analyte or Components) P2O7 43.5 to 48.5%
Chemical Name or Material Copper (II) pyrophosphate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'