Learn More
Copper(II) phthalocyanine, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Copper(II) phthalocyanine are involved in the study of photosensitizer chemistry for uniform polymerization, luminescence chemistry and spectrophotometric analysis, organic synthesis and polymerization. This pigment is used in enamels, linoleum, inks, plastics, and rubber goods. Photoisomerizable phthalocyanines are used in rewritable CD or DVD printing. Other applications in organic solar cells, biosensitizers and display devices such as OLED , OTFT, Wearable Display, and e-paper.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 350°C |
Color | Blue to Purple |
Quantity | 10 g |
Beilstein | 4121848 |
Merck Index | 14,2520 |
Solubility Information | It is insoluble in water. |
Formula Weight | 576.08 |
Physical Form | Powder |
Chemical Name or Material | Copper (II) phthalocyanine |
RUO – Research Use Only