missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog Number p-7048846
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
10294-46-9
Cl2CuH12O14
370.528
MFCD00149666
NHELIHXBJRANPL-UHFFFAOYSA-L
copper ii perchlorate hexahydrate, cupric perchlorate hexahydrate, unii-57jsh2lo1g, 57jsh2lo1g, copper diperchlorate hexahydrate, cupric diperchlorate hexahydrate, copper 2+ perchlorate hexahydrate, cupric perchlorate hexahydrate mi, acmc-20aldl, copper perchlorate cu clo4 2 hexahydrate
12846356
copper;diperchlorate;hexahydrate
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Cu+2]
This item is not returnable. View return policy
10294-46-9
Cl2CuH12O14
370.528
MFCD00149666
NHELIHXBJRANPL-UHFFFAOYSA-L
copper ii perchlorate hexahydrate, cupric perchlorate hexahydrate, unii-57jsh2lo1g, 57jsh2lo1g, copper diperchlorate hexahydrate, cupric diperchlorate hexahydrate, copper 2+ perchlorate hexahydrate, cupric perchlorate hexahydrate mi, acmc-20aldl, copper perchlorate cu clo4 2 hexahydrate
12846356
copper;diperchlorate;hexahydrate
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Cu+2]

Copper(II) perchlorate hexahydrate is a strong oxidizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 82°C
Physical Form Crystalline
Quantity 50 g
UN Number UN1481
Merck Index 14,2647
Solubility Information Soluble in water, methanol, ethanol, acetic acid, acetic anhydride, and acetone
Formula Weight 370.53 (262.44 Anhydrous)
Grade Reagent
Odor Odorless
Refractive Index 1.505
Sensitivity Hygroscopic
Density 2.23 g/mL
Chemical Name or Material Copper(II) perchlorate hexahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'