missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) perchlorate hexahydrate, 99.999% (metals basis), Thermo Scientific™

Catalog Number p-7059228
Click to view available options
Quantity:
50 g
This item is not returnable. View return policy
10294-46-9
Cl2CuH12O14
370.528
MFCD00149666
NHELIHXBJRANPL-UHFFFAOYSA-L
copper ii perchlorate hexahydrate, cupric perchlorate hexahydrate, unii-57jsh2lo1g, 57jsh2lo1g, copper diperchlorate hexahydrate, cupric diperchlorate hexahydrate, copper 2+ perchlorate hexahydrate, cupric perchlorate hexahydrate mi, acmc-20aldl, copper perchlorate cu clo4 2 hexahydrate
12846356
copper;diperchlorate;hexahydrate
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Cu+2]
This item is not returnable. View return policy
10294-46-9
Cl2CuH12O14
370.528
MFCD00149666
NHELIHXBJRANPL-UHFFFAOYSA-L
copper ii perchlorate hexahydrate, cupric perchlorate hexahydrate, unii-57jsh2lo1g, 57jsh2lo1g, copper diperchlorate hexahydrate, cupric diperchlorate hexahydrate, copper 2+ perchlorate hexahydrate, cupric perchlorate hexahydrate mi, acmc-20aldl, copper perchlorate cu clo4 2 hexahydrate
12846356
copper;diperchlorate;hexahydrate
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Cu+2]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 82°C
Physical Form Crystalline
Quantity 50 g
UN Number UN1481
Merck Index 14,2647
Solubility Information Soluble in water and ethanol. Very soluble in ethyl alcohol and acetone.
Formula Weight 370.53 (262.44 Anhydrous)
Percent Purity 99.999%
Odor Odorless
Refractive Index 1.505
Sensitivity Hygroscopic
Density 2.23 g/mL
Assay (metals basis)
Chemical Name or Material Copper (II) perchlorate hexahydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'