missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) oxide, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 100 g |
UN Number | UN3077 |
Formula Weight | 79.55g/mol |
Percent Purity | ≥99% (RT) |
Chemical Name or Material | Copper(II) oxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'