missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) oxide, 99+%, ACROS Organics™

Brand:  Acros Organics 221281000

Code : 29

Additional Details : CAS Number : 1317-38-0 Weight : 0.10000kg Transport : UN number : 3077 Chem class : 9 Pack group : III


Packaging Plastic bottle
Quantity 100g

 View more versions of this product

Product Code. 10750921

  • £38.80 / 100g

Estimated Shipment date
from Supplier 16-06-2020

Please to purchase this item. Need a web account? Register with us today!



Specifications

Chemical Identifiers

1317-38-0
79.54
QPLDLSVMHZLSFG-UHFFFAOYSA-N
14829
O=[Cu]
CuO
MFCD00010979
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
oxocopper

Specifications

Copper(II) oxide
1317-38-0
CuO
MFCD00010979
15, 2636
Solubility in water: insoluble. Other solubilities: soluble in dil.acids- slowly soluble in nh3,soluble in alkali cyanides and (nh4)2co3 solutions,practically insoluble in alcohol
O=[Cu]
79.54
79.54
99+%
50ppm max
Passes Test
500ppm max.
Plastic bottle
0.02% max.
1326.0°C
p.a.
99% min. (Complexometry)
CuO
04,108; 05,161
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
QPLDLSVMHZLSFG-UHFFFAOYSA-N
oxocopper
14829
Powder
500ppm max.
50ppm max.
0.02% max. (in dilute HCl)
0.002% max.
Solubility in water: insoluble. Other solubilities: soluble in dil.acids - slowly soluble in nh3, soluble in alkali cyanides and (nh4)2co3 solutions, practically insoluble in alcohol
Black
100g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Copper(II) oxide

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

SDS Documents