missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) oxalate hemihydrate, 98%, Thermo Scientific Chemicals

Catalog Number p-7022538
Click to view available options
Quantity:
50 g
250 g
5893-66-3
C4H8Cu2O10
343.19
MFCD00150451
NNZNHNMQOYUANB-UHFFFAOYSA-N
cupric oxalate, copper oxalate hemihydrate
122130000
copper;oxalic acid;hydrate
O.O.[Cu].[Cu].OC(=O)C(O)=O.OC(=O)C(O)=O
5893-66-3
C4H8Cu2O10
343.19
MFCD00150451
NNZNHNMQOYUANB-UHFFFAOYSA-N
cupric oxalate, copper oxalate hemihydrate
122130000
copper;oxalic acid;hydrate
O.O.[Cu].[Cu].OC(=O)C(O)=O.OC(=O)C(O)=O

Copper(II) oxalate hemihydrate is used as a catalyst for organic reactions; as stabilizer for acetylated polyformaldehyde; in anticaries compositions; in seed treatments to repel birds and rodents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) oxalate hemihydrate is used as a catalyst for organic reactions; as stabilizer for acetylated polyformaldehyde; in anticaries compositions; in seed treatments to repel birds and rodents.

Solubility
Soluble in NH{4}OH

Notes
Store in cool, dry, and well-ventilated area. Store away from oxidizing agents. Incompatible with acitve metals.
TRUSTED_SUSTAINABILITY
Assay Percent Range 98%
Quantity 50 g
UN Number UN3288
Merck Index 14,2645
Solubility Information Soluble in NH4OH
Formula Weight 160.57 (151.51 Anhydrous)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Copper(II) Oxalate Hemihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'