Learn More
Copper(II) nitrate hemi(pentahydrate), ACS, 98.0-102.0%, Thermo Scientific Chemicals
Descrizione
Catalyst, nitrating agent for aromatic organosilicon compounds, nickel-plating baths, coloring copper black and producing a burnished effect on ironCopper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Boiling Point | 170°C (decomposition) |
Melting Point | ∽115°C |
Physical Form | Crystalline Lump |
Quantity | 2 kg |
Assay Percent Range | 98.0-102.0% |
Solubility Information | Soluble in water and alcoholSoluble in water,ammonia and ethanol. |
Formula Weight | 232.59 (187.55anhydrous) |
Grade | ACS |
Sensitivity | Hygroscopic |
Density | 2.32 |
Vedi altri risultati |
RUO – Research Use Only