missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) nitrate, 2.5-hydrate, reagent ACS, +98%, ACROS Organics™

Brand:  Acros Organics 405851000

Code : 29

Additional Details : CAS Number : 19004-19-4 Weight : 0.15000kg Transport : UN number : 3260 Chem class : 8 Pack group : II


Quantity 100g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10413655

  • £11.90 / 100g


EU Stock 10
Add to basket


Specifications

Chemical Identifiers

Copper(II) nitrate, hemipentahydrate
ACS Reagent Grade
98+%
2·5H2O
MFCD02253001
19004-19-4
≥98%
2·5 H2O
232.6
Cupric nitrate

Specifications

Copper(II) nitrate, hemipentahydrate
ACS Reagent Grade
≥98%
2·5 H2O
232.6
17,366
Cupric nitrate
20ppm max.
50ppm max.
100ppm max.
50ppm max.
Solubility in water: soluble.
Blue
100g
19004-19-4
Crystalline Powder
98+%
2·5H2O
MFCD02253001
10, 2633
50ppm max.
0.01% max.
10ppm max.
Plastic bottle
100ppm max.
100ppm max.
114.0°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Copper(II) nitrate, 2.5-hydrate, reagent ACS

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS