missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) methoxide, typically 98% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7049706
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1184-54-9
MFCD00015611
This item is not returnable. View return policy
1184-54-9
MFCD00015611

Copper(II) methoxide is used as primary and secondary intermediates. Treatment of these methoxide derivatives with excess water results in the clean formation of mononuclear copper(II) hydroxide complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼206°C (decomposition)
Color Green
Odor Alcohol-like
Linear Formula Cu(OCH3)2
Quantity 1 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 125.61
Percent Purity 98% (typically)
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Copper(II) methoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'