missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Copper(II) fluoride hydrate, Puratronic™, 99.99% (metals basis)

Brand:  Alfa Aesar™ 013001.04

Code : NEW

Additional Details : CAS Number : 13454-88-1 Weight : 0.00200kg Transport : UN number : 3288 Chem class : 1 Pack group : III

Quantity 2g

 View more versions of this product

Product Code. 11362098

  • £22.00 / 2g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

Copper(II) fluoride hydrate
Puratronic™
MFCD00010977
14,2638
13454-88-1
101.54 (Anhydrous)
UN3288

Specifications

Copper(II) fluoride hydrate
Puratronic™
99.99%
101.54 (Anhydrous)
UN3288
2.934g/mL
Hygroscopic
>130°C (dec.)
13454-88-1
Crystalline
(metals basis)
MFCD00010977
14,2638
Odorless
Blue
2g
Videos
SDS