missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) fluoride, anhydrous, 99.5%, Thermo Scientific Chemicals

Catalog Number p-7048792
Click to view available options
Quantity:
10 g
50 g
7789-19-7
MFCD00010977
7789-19-7
MFCD00010977

Cathodes in nonaqueous galvanic cells, high temp. fluorinating agent

Cathodes in nonaqueous galvanic cells, high temp. fluorinating agent.Copper(II) fluoride is used in the preparation of fluorobenzene by reacting with benzene. It is also used in galvanic cells, high-temperature fluorinating agent and fluxing iron castings. Further, it is used as an efficient catalyst in Chan-Lam-Evans coupling reaction to synthesis the diaryl- ether compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 950°C (decomposition)
Color White
Physical Form Powder
Quantity 10 g
UN Number UN3260
Merck Index 14,2638
Solubility Information Solubility in water at 20° ≈ 4;7g/100mL
Formula Weight 101.54
Percent Purity 99.5%
Odor Odorless
Sensitivity Hygroscopic
Density 4.85 g/mL
Chemical Name or Material Copper(II) fluoride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'