missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Chloride Dihydrate, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7072942
Click to view available options
Quantity:
250 g
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
10125-13-0
Cl2CuH4O2
170.48
MFCD00149674
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
copper(2+) dihydrate dichloride
O.O.[Cl-].[Cl-].[Cu++]
This item is not returnable. View return policy
10125-13-0
Cl2CuH4O2
170.48
MFCD00149674
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
copper(2+) dihydrate dichloride
O.O.[Cl-].[Cl-].[Cu++]
Melting Point 598°C
Color Blue-Green
pH 3.0 to 3.8
Physical Form Solid
Quantity 250 g
Formula Weight 170.48g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(II) chloride dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'