missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) chloride dihydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog Number p-3593411
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10125-13-0
Cl2CuH4O2
170.48
MFCD00149674
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
61482
CHEBI:86318
O.O.[Cl-].[Cl-].[Cu++]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10125-13-0
Cl2CuH4O2
170.48
MFCD00149674
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
61482
CHEBI:86318
O.O.[Cl-].[Cl-].[Cu++]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 100.0°C
Color White
pH 3.0 to 3.8 (5% aq. soln. at 20°C)
Physical Form Crystals or Powder
Quantity 250 g
Assay Percent Range 99% min. (Iodometry)
Linear Formula CuCl2·2H2O
Fieser 01,163; 02,84; 03,66; 04,105; 05,158; 06,139; 07,79; 08,119; 09,123; 13,152
Merck Index 15, 2623
Solubility Information Solubility in water: 1150g/L (20°C). Other solubilities: soluble in alcohol, moderately soluble in acetone and ethylacetate, slightly soluble in ether
IUPAC Name dichlorocopper;dihydrate
Formula Weight 170.48
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Copper(II) chloride dihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'