missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Carbonate Basic, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7072941
Click to view available options
Quantity:
250 g
500 g
This item is not returnable. View return policy
12069-69-1
221.11
10976
ZMMDPCMYTCRWFF-UHFFFAOYSA-J
dicopper(2+) dihydroxide carbonate
[OH-].[OH-].[Cu++].[Cu++].[O-]C([O-])=O
This item is not returnable. View return policy
12069-69-1
221.11
10976
ZMMDPCMYTCRWFF-UHFFFAOYSA-J
dicopper(2+) dihydroxide carbonate
[OH-].[OH-].[Cu++].[Cu++].[O-]C([O-])=O

Copper(II) Carbonate Basic, Extra Pure, SLR

TRUSTED_SUSTAINABILITY
Melting Point 200°C
Color Green
pH 8 to 9
Physical Form Powder/Solid
Quantity 250 g
Molecular Formula CH2Cu2O5
Formula Weight 221.1g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Copper (II) carbonate basic
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'