missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(Ii) Acetate Monohydrate, Honeywell™
Click to view available options
Quantity:
500 g
1 kg
50 kg
6 x 1 kg
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Quantity | 500 g |
Linear Formula | Cu(CO2CH3)2 · H2O |
UN Number | 1759 |
Beilstein | 3730548 |
IUPAC Name | copper(2+) diacetate hydrate |
Formula Weight | 199.65g/mol |
Percent Purity | ≥99% (iodometric) |
Density | Vapor Density: 6.8 (vs air) |
Chemical Name or Material | Copper(Ii) Acetate Monohydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'