missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) acetate monohydrate, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Boiling Point | 240.0°C |
Melting Point | 115.0°C |
Color | Blue-Green or Green |
Physical Form | Solid |
Quantity | 500 g |
Linear Formula | (CH3CO2)2Cu·H2O |
Fieser | 01,159; 02,84; 03,65; 04,105; 05,156; 06,138; 10,103; 12,140; 14,199; 15,99; 17,176 |
Merck Index | 15,2614 |
Solubility Information | Solubility in water: 72g/L (20°C). Other solubilities: soluble in alcohol and ether |
Formula Weight | 199.65 |
Show More |
Safety and Handling
Product Identifier
- Copper(II) acetate monohydrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'