missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) acetate monohydrate, 98+%, Thermo Scientific Chemicals

Catalog Number p-7023122
Click to view available options
Quantity:
250 g
1000 g
5000 g
6046-93-1
C4H8CuO5
199.65
MFCD00149570
NWFNSTOSIVLCJA-UHFFFAOYSA-L
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
165397
copper;diacetate;hydrate
O.[Cu++].CC([O-])=O.CC([O-])=O
6046-93-1
C4H8CuO5
199.65
MFCD00149570
NWFNSTOSIVLCJA-UHFFFAOYSA-L
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
165397
copper;diacetate;hydrate
O.[Cu++].CC([O-])=O.CC([O-])=O

Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.

Solubility
Soluble in water and alcohol. Slightly soluble in ether and glycerine.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 115°C
Quantity 250 g
Linear Formula Cu(OOCCH3)2·H2O
UN Number UN3077
Merck Index 14,2624
Solubility Information Soluble in water and alcohol. Slightly soluble in ether and glycerine.
Formula Weight 199.65 (181.64 Anhydrous)
Percent Purity ≥98%
Density 1.882 g/mL
Chemical Name or Material Copper(II) acetate monohydrate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(II) acetate monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Copper diacetate monohydrate, Acetic acid, copper(2+) salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'