missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Copper(I) trifluoromethanesulfonate benzene complex (2:1)

Brand:  Alfa Aesar™ 016986.06

Code : NEW

Additional Details : CAS Number : 37234-97-2 Weight : 0.00500kg Transport : UN number : 3181 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 11363317

  • £183.30 / 5g


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Specifications

Chemical Identifiers

Copper(I) trifluoromethanesulfonate benzene complex (2:1)
503.33
UN3181
37234-97-2
MFCD00043300
Copper(I) triflate; Bis[copper(I) trifluoromethanesulfonate]

Specifications

Copper(I) trifluoromethanesulfonate benzene complex (2:1)
5g
Powder
MFCD00043300
Copper(I) triflate; Bis[copper(I) trifluoromethanesulfonate]
37234-97-2
Air & Moisture Sensitive
503.33
UN3181
Soluble in hot benzene
Videos
SDS